gRNA Design Details

Target DNA Sequence
GCTTTGATGGATGCGCTCTG
PAM Sequence
CGG
Target locus
Chr.11: 104163833 - 104163855 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 91.71/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
PDGFD
Entrez Gene ID
80310
Gene Name
platelet derived growth factor D
Gene Aliases
IEGF, MSTP036, SCDGF-B, SCDGFB
Chromosome Location
Chr.11: 103907186 - 104164299 on GRCh38
UniGene ID
Hs.352298
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_025208.4 NP_079484.1 1
NM_033135.3 NP_149126.1 1