gRNA Design Details

Target DNA Sequence
GAGAAGTTTTTATGCTGCCA
PAM Sequence
GGG
Target locus
Chr.6: 71293328 - 71293350 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 77.81/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
OGFRL1
Entrez Gene ID
79627
Gene Name
opioid growth factor receptor like 1
Gene Aliases
dJ331H24.1
Chromosome Location
Chr.6: 71288296 - 71309059 on GRCh38
UniGene ID
Hs.648434
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001324266.1 NP_001311195.1 2
NM_024576.4 NP_078852.3 2
XM_011536117.2 XP_011534419.1 2
XM_011536118.2 XP_011534420.1 2
XM_017011303.1 XP_016866792.1 2
XM_017011304.1 XP_016866793.1 2
XM_017011305.1 XP_016866794.1 2
XM_017011306.1 XP_016866795.1 2