gRNA Design Details
- Target DNA Sequence
- TGCCCTGATGATCAGATCAG
- PAM Sequence
- AGG
- Target locus
-
Chr.12: 4372674 - 4372696 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
87.83/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- FGF23
- Entrez Gene ID
-
8074
- Gene Name
- fibroblast growth factor 23
- Gene Aliases
-
ADHR, FGFN, HPDR2, HYPF, PHPTC
- Chromosome Location
-
Chr.12: 4368227 - 4379728 on GRCh38
- UniGene ID
- Hs.287370
- Species
-
Human