gRNA Design Details

Target DNA Sequence
GACCTCCAGCAACGATGATG
PAM Sequence
AGG
Target locus
Chr.5: 177090752 - 177090774 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 89.51/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
FGFR4
Entrez Gene ID
2264
Gene Name
fibroblast growth factor receptor 4
Gene Aliases
CD334, JTK2, TKF
Chromosome Location
Chr.5: 177086872 - 177098144 on GRCh38
UniGene ID
Hs.165950
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001291980.1 NP_001278909.1 4
NM_002011.4 NP_002002.3 4
NM_022963.3 NP_075252.2 3
NM_213647.2 NP_998812.1 4
XM_005265838.3 XP_005265895.1 4
XM_011534464.2 XP_011532766.1 4
XM_011534465.2 XP_011532767.1 4