gRNA Design Details

Target DNA Sequence
TTGAGACGGACGATCACGGA
PAM Sequence
GGG
Target locus
Chr.5: 100535802 - 100535824 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 95.91/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
FAM174A
Entrez Gene ID
345757
Gene Name
family with sequence similarity 174 member A
Gene Aliases
HGS_RE408, TMEM157, UNQ1912
Chromosome Location
Chr.5: 100535305 - 100586741 on GRCh38
UniGene ID
Hs.356108
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_198507.2 NP_940909.1 1
XM_006714600.1 XP_006714663.1 1