gRNA Design Details

Target DNA Sequence
AATATAGCTGTCCAAGTGCG
PAM Sequence
AGG
Target locus
Chr.9: 38413274 - 38413296 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 93.83/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
IGFBPL1
Entrez Gene ID
347252
Gene Name
insulin like growth factor binding protein like 1
Gene Aliases
IGFBP-RP4, IGFBPRP4, bA113O24.1
Chromosome Location
Chr.9: 38406528 - 38424447 on GRCh38
UniGene ID
Hs.349705
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001007563.2 NP_001007564.1 3