gRNA Design Details
- Target DNA Sequence
- GAACAAAATTCCATTTAAGC
- PAM Sequence
- CGG
- Target locus
-
Chr.1: 23378729 - 23378751 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
75.16/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Ogfrl1
- Entrez Gene ID
-
70155
- Gene Name
- opioid growth factor receptor-like 1
- Gene Aliases
-
2210417C17Rik
- Chromosome Location
-
Chr.1: 23366424 - 23383210 on GRCm38
- UniGene ID
- Mm.28013
- Species
-
Mouse