gRNA Design Details
- Target DNA Sequence
- CAGCAAAGGCGCCGACTATA
- PAM Sequence
- TGG
- Target locus
-
Chr.18: 3414189 - 3414211 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
84.33/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Cul2
- Entrez Gene ID
-
71745
- Gene Name
- cullin 2
- Gene Aliases
-
1300003D18Rik, 4932411N15Rik, AI327301, mKIAA4106
- Chromosome Location
-
Chr.18: 3382975 - 3436700 on GRCm38
- UniGene ID
- Mm.291707
- Species
-
Mouse