gRNA Design Details

Target DNA Sequence
AGCAGCTAAAGTTCGCGCAC
PAM Sequence
AGG
Target locus
Chr.9: 6168824 - 6168846 on GRCm38
Strand
Reverse
Application
Gene Knock-Out
Score: 93.18/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Pdgfd
Entrez Gene ID
71785
Gene Name
platelet-derived growth factor, D polypeptide
Gene Aliases
1110003I09Rik
Chromosome Location
Chr.9: 6168362 - 6536014 on GRCm38
UniGene ID
Mm.390122
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_027924.2 NP_082200.1 1