gRNA Design Details
- Target DNA Sequence
- TGACCATCAATTCGGACTAG
- PAM Sequence
- AGG
- Target locus
-
Chr.9: 6288632 - 6288654 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
92.36/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Pdgfd
- Entrez Gene ID
-
71785
- Gene Name
- platelet-derived growth factor, D polypeptide
- Gene Aliases
-
1110003I09Rik
- Chromosome Location
-
Chr.9: 6168362 - 6536014 on GRCm38
- UniGene ID
- Mm.390122
- Species
-
Mouse