gRNA Design Details
- Target DNA Sequence
- CGACTTGCAGCTCGCTCACT
- PAM Sequence
- AGG
- Target locus
-
Chr.4: 141793899 - 141793921 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
85.38/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Casp9
- Entrez Gene ID
-
12371
- Gene Name
- caspase 9
- Gene Aliases
-
AI115399, APAF-3, AW493809, CASP-9, Caspase-9, ICE-LAP6, Mch6
- Chromosome Location
-
Chr.4: 141793612 - 141815978 on GRCm38
- UniGene ID
- Mm.88829
- Species
-
Mouse