gRNA Design Details
- Target DNA Sequence
- TTATGCTCAGAGCTGAGGGG
- PAM Sequence
- AGG
- Target locus
-
Chr.5: 137031085 - 137031107 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
86.42/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Vgf
- Entrez Gene ID
-
381677
- Gene Name
- VGF nerve growth factor inducible
- Gene Aliases
-
Gm1052
- Chromosome Location
-
Chr.5: 137025302 - 137033351 on GRCm38
- UniGene ID
- Mm.389697
- Species
-
Mouse