gRNA Design Details
- Target DNA Sequence
- TTTGGTGGAGTCCAATTCGG
- PAM Sequence
- TGG
- Target locus
-
Chr.4: 9844581 - 9844603 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
92.23/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Gdf6
- Entrez Gene ID
-
242316
- Gene Name
- growth differentiation factor 6
- Gene Aliases
-
BMP-13, BMP13, GDF16
- Chromosome Location
-
Chr.4: 9844372 - 9862345 on GRCm38
- UniGene ID
- Mm.302555
- Species
-
Mouse