gRNA Design Details
- Target DNA Sequence
- GGCATTGATGCCCAGCTTCT
- PAM Sequence
- CGG
- Target locus
-
Chr.4: 9844809 - 9844831 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
84.76/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Gdf6
- Entrez Gene ID
-
242316
- Gene Name
- growth differentiation factor 6
- Gene Aliases
-
BMP-13, BMP13, GDF16
- Chromosome Location
-
Chr.4: 9844372 - 9862345 on GRCm38
- UniGene ID
- Mm.302555
- Species
-
Mouse