gRNA Design Details

Target DNA Sequence
GGATGGCGACATGAAGCAGG
PAM Sequence
AGG
Target locus
Chr.5: 16553987 - 16554009 on GRCm38
Strand
Reverse
Application
Gene Knockout
Score: 83.77/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Hgf
Entrez Gene ID
15234
Gene Name
hepatocyte growth factor
Gene Aliases
C230052L06Rik, HGF/SF, NK1, NK2, SF, SF/HGF
Chromosome Location
Chr.5: 16553335 - 16622408 on GRCm38
UniGene ID
Mm.267078
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001289458.1 NP_001276387.1 2
NM_001289459.1 NP_001276388.1 2
NM_001289460.1 NP_001276389.1 2
NM_001289461.1 NP_001276390.1 1
NM_010427.5 NP_034557.3 2