gRNA Design Details

Target DNA Sequence
TCAGACCTAGATACCACCCT
PAM Sequence
CGG
Application
Gene Knock-Out
Score: 86.56/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Fgf13
Entrez Gene ID
14168
Gene Name
fibroblast growth factor 13
Gene Aliases
Fhf2
Chromosome Location
Chr.X: 59062139 - 59585572 on GRCm38
UniGene ID
Mm.7995
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001290414.1 NP_001277343.1 6
NM_001290415.1 NP_001277344.1 4
NM_010200.3 NP_034330.2 4