gRNA Design Details

Target DNA Sequence
ATACAAGTTTGAGAGCTGGG
PAM Sequence
GGG
Target locus
Chr.2: 91930947 - 91930969 on GRCm38
Strand
Reverse
Application
Gene Knock-Out
Score: 84.47/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Mdk
Entrez Gene ID
17242
Gene Name
midkine
Gene Aliases
MK, Mek
Chromosome Location
Chr.2: 91929805 - 91932297 on GRCm38
UniGene ID
Mm.906
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001012335.2 NP_001012335.1 4
NM_001012336.2 NP_001012336.1 4
NM_001291481.1 NP_001278410.1 4
NM_001291482.1 NP_001278411.1 3
NM_010784.5 NP_034914.1 4