gRNA Design Details
- Target DNA Sequence
- GTAGTAGCAAGTGTACCATC
- PAM Sequence
- CGG
- Target locus
-
Chr.5: 75163769 - 75163791 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
88.25/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Pdgfra
- Entrez Gene ID
-
18595
- Gene Name
- platelet derived growth factor receptor, alpha polypeptide
- Gene Aliases
-
AI115593, CD140a, Pdgfr-2
- Chromosome Location
-
Chr.5: 75151401 - 75198211 on GRCm38
- UniGene ID
- Mm.221403
- Species
-
Mouse