gRNA Design Details
- Target DNA Sequence
- TTCACCTACGCTTCCCAAGG
- PAM Sequence
- AGG
- Target locus
-
Chr.17: 56182675 - 56182697 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
96.18/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Mydgf
- Entrez Gene ID
-
28106
- Gene Name
- myeloid derived growth factor
- Gene Aliases
-
D17Wsu104e, Il25, Ly6elg
- Chromosome Location
-
Chr.17: 56176541 - 56183920 on GRCm38
- UniGene ID
- Mm.294870
- Species
-
Mouse