gRNA Design Details
- Target DNA Sequence
- GCTTCGAGTCTGAGAAAAGG
- PAM Sequence
- CGG
- Target locus
-
Chr.15: 102407865 - 102407887 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
85.46/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Sp1
- Entrez Gene ID
-
20683
- Gene Name
- trans-acting transcription factor 1
- Gene Aliases
-
1110003E12Rik, AA450830, AI845540, Sp1-1
- Chromosome Location
-
Chr.15: 102406316 - 102436404 on GRCm38
- UniGene ID
- Mm.4618
- Species
-
Mouse