gRNA Design Details
- Target DNA Sequence
- TCGTATAACGGATTTCCACC
- PAM Sequence
- GGG
- Target locus
-
Chr.8: 72400208 - 72400230 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
89.91/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Eps15l1
- Entrez Gene ID
-
13859
- Gene Name
- epidermal growth factor receptor pathway substrate 15-like 1
- Gene Aliases
-
9830147J04Rik, AI593686, Eps15-rs, Eps15R
- Chromosome Location
-
Chr.8: 72340990 - 72421565 on GRCm38
- UniGene ID
- Mm.288894
- Species
-
Mouse