gRNA Design Details
- Target DNA Sequence
- TTGGCAGACCCCGAAGGTAA
- PAM Sequence
- AGG
- Target locus
-
Chr.8: 72399022 - 72399044 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
87.75/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Eps15l1
- Entrez Gene ID
-
13859
- Gene Name
- epidermal growth factor receptor pathway substrate 15-like 1
- Gene Aliases
-
9830147J04Rik, AI593686, Eps15-rs, Eps15R
- Chromosome Location
-
Chr.8: 72340990 - 72421565 on GRCm38
- UniGene ID
- Mm.288894
- Species
-
Mouse