gRNA Design Details
- Target DNA Sequence
- GCAAGTCTCAGTCGGCCAAC
- PAM Sequence
- TGG
- Target locus
-
Chr.7: 45109143 - 45109165 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
70.99/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- TBRG4
- Entrez Gene ID
-
9238
- Gene Name
- transforming growth factor beta regulator 4
- Gene Aliases
-
CPR2, FASTKD4
- Chromosome Location
-
Chr.7: 45100100 - 45111747 on GRCh38
- UniGene ID
- Hs.231411
- Species
-
Human