gRNA Design Details
- Target DNA Sequence
- TGGCTTTATCGAGATCATCT
- PAM Sequence
- TGG
- Target locus
-
Chr.11: 106117510 - 106117532 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
84.29/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Map3k3
- Entrez Gene ID
-
26406
- Gene Name
- mitogen-activated protein kinase kinase kinase 3
- Gene Aliases
-
AW548911, MAPKKK3, Mekk3, mKIAA4031
- Chromosome Location
-
Chr.11: 106084902 - 106156656 on GRCm38
- UniGene ID
- Mm.27041
- Species
-
Mouse