gRNA Design Details
- Target DNA Sequence
- GCAGCTCCTCATCGTGTTGG
- PAM Sequence
- TGG
- Target locus
-
Chr.4: 47353344 - 47353366 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
89.8/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Tgfbr1
- Entrez Gene ID
-
21812
- Gene Name
- transforming growth factor, beta receptor I
- Gene Aliases
-
ALK5, AU017191, Alk-5, TbetaR-I, TbetaRI
- Chromosome Location
-
Chr.4: 47353113 - 47414926 on GRCm38
- UniGene ID
- Mm.197552
- Species
-
Mouse