gRNA Design Details
- Target DNA Sequence
- GGGAGCCGAAATCGCAGAAG
- PAM Sequence
- AGG
- Target locus
-
Chr.10: 24596046 - 24596068 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
88.19/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Ctgf
- Entrez Gene ID
-
14219
- Gene Name
- connective tissue growth factor
- Gene Aliases
-
Ccn2, Fisp12, Hcs24, fisp-12
- Chromosome Location
-
Chr.10: 24595442 - 24598683 on GRCm38
- UniGene ID
- Mm.390287
- Species
-
Mouse