gRNA Design Details
- Target DNA Sequence
- AGCCGATCGGTTTCAGTTGC
- PAM Sequence
- TGG
- Target locus
-
Chr.11: 49854338 - 49854360 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
91.82/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Mapk9
- Entrez Gene ID
-
26420
- Gene Name
- mitogen-activated protein kinase 9
- Gene Aliases
-
AI851083, JNK2, Prkm9, p54aSAPK
- Chromosome Location
-
Chr.11: 49846751 - 49886421 on GRCm38
- UniGene ID
- Mm.68933
- Species
-
Mouse