gRNA Design Details

Target DNA Sequence
AATGGATGCTAACTTATGTC
PAM Sequence
AGG
Target locus
Chr.5: 180261782 - 180261804 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 61.57/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
MAPK9
Entrez Gene ID
5601
Gene Name
mitogen-activated protein kinase 9
Gene Aliases
JNK-55, JNK2, JNK2A, JNK2ALPHA, JNK2B, JNK2BETA, PRKM9, SAPK, SAPK1a, p54a, p54aSAPK
Chromosome Location
Chr.5: 180233594 - 180292071 on GRCh38
UniGene ID
Hs.484371
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001135044.1 NP_001128516.1 5
NM_001308244.1 NP_001295173.1 4
NM_002752.4 NP_002743.3 5
NM_139068.2 NP_620707.1 5
NM_139069.2 NP_620708.1 5
NM_139070.2 NP_620709.1 5
XM_005265940.4 XP_005265997.1 6
XM_006714891.2 XP_006714954.1 6
XM_017009638.1 XP_016865127.1 6
XM_017009639.1 XP_016865128.1 7
XM_017009640.1 XP_016865129.1 6
XM_017009641.1 XP_016865130.1 6
XM_017009642.1 XP_016865131.1 5
XM_017009643.1 XP_016865132.1 5