Targeted Isoforms | Translated Protein | Targeted Exon(s) | |
---|---|---|---|
RefSeq | NM_001321931.1 | NP_001308860.1 | 3 |
NM_001321932.1 | NP_001308861.1 | 3 | |
NM_001321933.1 | NP_001308862.1 | 3 | |
NM_001321934.1 | NP_001308863.1 | 2 | |
NM_001321935.1 | NP_001308864.1 | 2 | |
NM_001321936.1 | NP_001308865.1 | 3 | |
NM_001321937.1 | NP_001308866.1 | 2 | |
NM_001321938.1 | NP_001308867.1 | 4 | |
NM_001321940.1 | NP_001308869.1 | 3 | |
NM_001321941.1 | NP_001308870.1 | 3 | |
NM_001321942.1 | NP_001308871.1 | 3 | |
NM_001321943.1 | NP_001308872.1 | 3 | |
NM_001321944.1 | NP_001308873.1 | 3 | |
NM_001321945.1 | NP_001308874.1 | 3 | |
NM_001321946.1 | NP_001308875.1 | 4 | |
NM_001321947.1 | NP_001308876.1 | 3 | |
NM_001321948.1 | NP_001308877.1 | 3 | |
NM_001321949.1 | NP_001308878.1 | 3 | |
NM_004115.3 | NP_004106.1 | 2 | |
NM_175929.2 | NP_787125.1 | 2 |