gRNA Design Details
- Target DNA Sequence
- GACAGAGAACTGCCGCTCAG
- PAM Sequence
- TGG
- Target locus
-
Chr.17: 12748644 - 12748666 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
82.93/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Igf2r
- Entrez Gene ID
-
16004
- Gene Name
- insulin-like growth factor 2 receptor
- Gene Aliases
-
AI661837, CD222, CI-MPR, M6P/IGF2R, Mpr300
- Chromosome Location
-
Chr.17: 12682406 - 12769706 on GRCm38
- UniGene ID
- Mm.26553
- Species
-
Mouse