gRNA Design Details
- Target DNA Sequence
- GCCCGAATTCATGCCTGAGC
- PAM Sequence
- TGG
- Target locus
-
Chr.13: 26769698 - 26769720 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
87.78/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Hdgfl1
- Entrez Gene ID
-
15192
- Gene Name
- hepatoma derived growth factor-like 1
- Gene Aliases
-
4921520N01Rik, HRP-1, Hdgfrp1, Pwwp1
- Chromosome Location
-
Chr.13: 26768172 - 26770165 on GRCm38
- UniGene ID
- Mm.7411
- Species
-
Mouse