gRNA Design Details
- Target DNA Sequence
- ACACTCGTAGCCGTTTGCTC
- PAM Sequence
- CGG
- Target locus
-
Chr.2: 126035823 - 126035845 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
91.82/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Fgf7
- Entrez Gene ID
-
14178
- Gene Name
- fibroblast growth factor 7
- Gene Aliases
-
Kgf
- Chromosome Location
-
Chr.2: 126001108 - 126091185 on GRCm38
- UniGene ID
- Mm.330557
- Species
-
Mouse