gRNA Design Details
- Target DNA Sequence
- ACCGGAGGAACCACCAAGAA
- PAM Sequence
- TGG
- Target locus
-
Chr.11: 98451117 - 98451139 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
83.25/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Grb7
- Entrez Gene ID
-
14786
- Gene Name
- growth factor receptor bound protein 7
- Gene Aliases
-
mKIAA4028
- Chromosome Location
-
Chr.11: 98446834 - 98455373 on GRCm38
- UniGene ID
- Mm.276702
- Species
-
Mouse