gRNA Design Details
- Target DNA Sequence
- GCCCGGACTGTAACCCCAAA
- PAM Sequence
- AGG
- Target locus
-
Chr.2: 155944697 - 155944719 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
85.82/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Gdf5
- Entrez Gene ID
-
14563
- Gene Name
- growth differentiation factor 5
- Gene Aliases
-
BMP-14, Cdmp-1, bp, brp
- Chromosome Location
-
Chr.2: 155941023 - 155945364 on GRCm38
- UniGene ID
- Mm.4744
- Species
-
Mouse