gRNA Design Details
- Target DNA Sequence
- GACCCAACCTTCAGTGGTGT
- PAM Sequence
- TGG
- Target locus
-
Chr.1: 60912433 - 60912455 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
86.91/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Ctla4
- Entrez Gene ID
-
12477
- Gene Name
- cytotoxic T-lymphocyte-associated protein 4
- Gene Aliases
-
Cd152, Ctla-4, Ly-56
- Chromosome Location
-
Chr.1: 60909025 - 60915832 on GRCm38
- UniGene ID
- Mm.390
- Species
-
Mouse