gRNA Design Details
- Target DNA Sequence
- TTCTGGAAGCAGGTGCTGGG
- PAM Sequence
- AGG
- Target locus
-
Chr.10: 91908623 - 91908645 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
67.22/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- FGFBP3
- Entrez Gene ID
-
143282
- Gene Name
- fibroblast growth factor binding protein 3
- Gene Aliases
-
C10orf13, FGF-BP3
- Chromosome Location
-
Chr.10: 91906588 - 91909501 on GRCh38
- UniGene ID
- Hs.466120
- Species
-
Human