gRNA Design Details

Target DNA Sequence
TCGCCACGTGTGTGAAATGC
PAM Sequence
TGG
Target locus
Chr.17: 39742963 - 39742985 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 87.07/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
GRB7
Entrez Gene ID
2886
Gene Name
growth factor receptor bound protein 7
Gene Aliases
-
Chromosome Location
Chr.17: 39737909 - 39747285 on GRCh38
UniGene ID
Hs.86859
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001030002.2 NP_001025173.1 4
NM_001242442.1 NP_001229371.1 4
NM_001242443.1 NP_001229372.1 4
NM_005310.3 NP_005301.2 4
XM_017024536.1 XP_016880025.1 3
XM_017024537.1 XP_016880026.1 4
XM_017024538.1 XP_016880027.1 3