gRNA Design Details

Target DNA Sequence
GACGTATTCCAACATGACCA
PAM Sequence
CGG
Target locus
Chr.10: 35071228 - 35071250 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 92.76/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
CUL2
Entrez Gene ID
8453
Gene Name
cullin 2
Gene Aliases
-
Chromosome Location
Chr.10: 35008551 - 35126899 on GRCh38
UniGene ID
Hs.82919
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001198777.1 NP_001185706.1 2
NM_001198778.1 NP_001185707.1 2
NM_001198779.1 NP_001185708.1 2
NM_001324375.1 NP_001311304.1 2
NM_001324376.1 NP_001311305.1 2
NM_003591.3 NP_003582.2 2
XM_011519743.1 XP_011518045.1 4
XM_011519744.1 XP_011518046.1 3
XM_011519745.1 XP_011518047.1 3
XM_011519747.1 XP_011518049.1 3