gRNA Design Details

Target DNA Sequence
CGATGCCCACTCTGCAGTAG
PAM Sequence
AGG
Target locus
Chr.4: 80267094 - 80267116 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 83.56/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
FGF5
Entrez Gene ID
2250
Gene Name
fibroblast growth factor 5
Gene Aliases
HBGF-5, Smag-82, TCMGLY
Chromosome Location
Chr.4: 80266588 - 80291017 on GRCh38
UniGene ID
Hs.37055
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_004464.3 NP_004455.2 1
NM_033143.2 NP_149134.1 1