gRNA Design Details

Target DNA Sequence
CATACAAACGGCCTATCTCG
PAM Sequence
AGG
Target locus
Chr.9: 99128992 - 99129014 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 96.18/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
TGFBR1
Entrez Gene ID
7046
Gene Name
transforming growth factor beta receptor 1
Gene Aliases
AAT5, ACVRLK4, ALK-5, ALK5, ESS1, LDS1, LDS1A, LDS2A, MSSE, SKR4, TGFR-1, tbetaR-I
Chromosome Location
Chr.9: 99104038 - 99154192 on GRCh38
UniGene ID
Hs.494622
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001130916.2 NP_001124388.1 2
NM_001306210.1 NP_001293139.1 2
NM_004612.3 NP_004603.1 2
XM_011518948.2 XP_011517250.1 2
XM_011518949.2 XP_011517251.1 2
XM_011518950.2 XP_011517252.1 2
XM_017015063.1 XP_016870552.1 2