gRNA Design Details
- Target DNA Sequence
- GAACTGGCAGCTGTCATTGC
- PAM Sequence
- TGG
- Target locus
-
Chr.9: 99132538 - 99132560 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
84.43/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- TGFBR1
- Entrez Gene ID
-
7046
- Gene Name
- transforming growth factor beta receptor 1
- Gene Aliases
-
AAT5, ACVRLK4, ALK-5, ALK5, ESS1, LDS1, LDS1A, LDS2A, MSSE, SKR4, TGFR-1, tbetaR-I
- Chromosome Location
-
Chr.9: 99104038 - 99154192 on GRCh38
- UniGene ID
- Hs.494622
- Species
-
Human