gRNA Design Details
- Target DNA Sequence
- ACATCGGCGGCCACTCTCAA
- PAM Sequence
- AGG
- Target locus
-
Chr.6: 41775432 - 41775454 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
90.7/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- FRS3
- Entrez Gene ID
-
10817
- Gene Name
- fibroblast growth factor receptor substrate 3
- Gene Aliases
-
FRS2-beta, FRS2B, FRS2beta, SNT-2, SNT2
- Chromosome Location
-
Chr.6: 41770176 - 41779905 on GRCh38
- UniGene ID
- Hs.194208
- Species
-
Human