gRNA Design Details

Target DNA Sequence
ACATCGGCGGCCACTCTCAA
PAM Sequence
AGG
Target locus
Chr.6: 41775432 - 41775454 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 90.7/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
FRS3
Entrez Gene ID
10817
Gene Name
fibroblast growth factor receptor substrate 3
Gene Aliases
FRS2-beta, FRS2B, FRS2beta, SNT-2, SNT2
Chromosome Location
Chr.6: 41770176 - 41779905 on GRCh38
UniGene ID
Hs.194208
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_006653.4 NP_006644.1 4
XM_011514254.2 XP_011512556.1 4
XM_011514255.2 XP_011512557.1 4
XM_017010193.1 XP_016865682.1 4