gRNA Design Details
- Target DNA Sequence
- ATCATTAGGGAGGAAGCCCA
- PAM Sequence
- CGG
- Target locus
-
Chr.5: 150134988 - 150135010 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
90.47/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PDGFRB
- Entrez Gene ID
-
5159
- Gene Name
- platelet derived growth factor receptor beta
- Gene Aliases
-
CD140B, IBGC4, IMF1, JTK12, KOGS, PDGFR, PDGFR-1, PDGFR1, PENTT
- Chromosome Location
-
Chr.5: 150113839 - 150155884 on GRCh38
- UniGene ID
- Hs.509067
- Species
-
Human