gRNA Design Details
- Target DNA Sequence
- CCGTGCAGATGGAATCATCT
- PAM Sequence
- AGG
- Target locus
-
Chr.2: 203870767 - 203870789 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
87.37/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- CTLA4
- Entrez Gene ID
-
1493
- Gene Name
- cytotoxic T-lymphocyte associated protein 4
- Gene Aliases
-
ALPS5, CD, CD152, CELIAC3, CTLA-4, GRD4, GSE, IDDM12
- Chromosome Location
-
Chr.2: 203867788 - 203873960 on GRCh38
- UniGene ID
- Hs.247824
- Species
-
Human