gRNA Design Details

Target DNA Sequence
TTCACTTACGCCTCTCAAGG
PAM Sequence
AGG
Target locus
Chr.19: 4668605 - 4668627 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 89.08/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
MYDGF
Entrez Gene ID
56005
Gene Name
myeloid derived growth factor
Gene Aliases
C19orf10, EUROIMAGE1875335, IL25, IL27, IL27w, R33729_1, SF20
Chromosome Location
Chr.19: 4657545 - 4670403 on GRCh38
UniGene ID
Hs.465645
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_019107.3 NP_061980.1 2