gRNA Design Details
- Target DNA Sequence
- GTTCCCGTACAAGGAGTGCA
- PAM Sequence
- AGG
- Target locus
-
Chr.6: 22569743 - 22569765 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
91.84/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- HDGFL1
- Entrez Gene ID
-
154150
- Gene Name
- hepatoma derived growth factor-like 1
- Gene Aliases
-
PWWP1, dJ309H15.1
- Chromosome Location
-
Chr.6: 22569449 - 22570521 on GRCh38
- UniGene ID
- Hs.629246
- Species
-
Human