gRNA Design Details

Target DNA Sequence
GTTCCCGTACAAGGAGTGCA
PAM Sequence
AGG
Target locus
Chr.6: 22569743 - 22569765 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 91.84/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
HDGFL1
Entrez Gene ID
154150
Gene Name
hepatoma derived growth factor-like 1
Gene Aliases
PWWP1, dJ309H15.1
Chromosome Location
Chr.6: 22569449 - 22570521 on GRCh38
UniGene ID
Hs.629246
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_138574.2 NP_612641.2 1