gRNA Design Details
- Target DNA Sequence
- GATACGACCGATGAGGACAC
- PAM Sequence
- GGG
- Target locus
-
Chr.2: 180589553 - 180589575 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
89.66/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Ogfr
- Entrez Gene ID
-
72075
- Gene Name
- opioid growth factor receptor
- Gene Aliases
-
2010013E17Rik
- Chromosome Location
-
Chr.2: 180589407 - 180595837 on GRCm38
- UniGene ID
- Mm.250418
- Species
-
Mouse