gRNA Design Details
- Target DNA Sequence
- ATATGCTGCAGCAAGGACCA
- PAM Sequence
- AGG
- Target locus
-
Chr.5: 132864376 - 132864398 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
88.98/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GDF9
- Entrez Gene ID
-
2661
- Gene Name
- growth differentiation factor 9
- Gene Aliases
-
-
- Chromosome Location
-
Chr.5: 132861181 - 132866884 on GRCh38
- UniGene ID
- Hs.25022
- Species
-
Human